Key Documents
530662
Sigma-Aldrich
2,6-Diamino-8-purinol hemisulfate salt hydrate
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(C5H6N6O)2 · H2SO4 · xH2O
CAS Number:
Molecular Weight:
332.28 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
mp
324-329 °C (lit.)
SMILES string
O.OS(O)(=O)=O.Nc1nc(N)c2[nH]c(O)nc2n1.Nc3nc(N)c4[nH]c(O)nc4n3
Choose from one of the most recent versions:
Certificates of Analysis (COA)