Key Documents
529893
Sigma-Aldrich
(E)-1-Heptene-1,2-diboronic acid bis(pinacol) ester
95%
Synonym(s):
1-[cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)] heptene, 2,2′-[(1Z)-1-Pentyl-1,2-ethenediyl]bis[4,4,5,5-tetramethyl-1,3,2-dioxaborolane]
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C19H36B2O4
CAS Number:
Molecular Weight:
350.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
assay
95%
refractive index
n20/D 1.457 (lit.)
bp
330 °C (lit.)
density
0.949 g/mL at 25 °C (lit.)
SMILES string
CCCCC\C(=C\B1OC(C)(C)C(C)(C)O1)B2OC(C)(C)C(C)(C)O2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)