Key Documents
528528
Sigma-Aldrich
3-Methoxy-2-naphthalenemethanol
97%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3OC10H6CH2OH
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
mp
70-74 °C (lit.)
SMILES string
COc1cc2ccccc2cc1CO
InChI
1S/C12H12O2/c1-14-12-7-10-5-3-2-4-9(10)6-11(12)8-13/h2-7,13H,8H2,1H3
InChI key
QUFXVYRFRKHGBW-UHFFFAOYSA-N
Application
3-Methoxy-2-naphthalenemethanol may be used to synthesize 2-carbomethoxy-4-methoxy-6,7-benzoindole.[1]
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Rob C Oslund et al.
Journal of medicinal chemistry, 51(15), 4708-4714 (2008-07-09)
We report a series of inhibitors of secreted phospholipases A2 (sPLA2s) based on substituted indoles, 6,7-benzoindoles, and indolizines derived from LY315920, a well-known indole-based sPLA2 inhibitor. Using the human group X sPLA2 crystal structure, we prepared a highly potent and
Questions
Reviews
No rating value
Active Filters