524484
Sulfuric acid-d2 solution
95-97 wt. % in D2O, 99 atom % D
Synonym(s):
Dideuterosulfuric acid solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
D2SO4/D2O
CAS Number:
Molecular Weight:
100.09
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
isotopic purity
99 atom % D
Quality Level
concentration
95-97 wt. % in D2O
refractive index
n20/D 1.423
mass shift
M+2
SMILES string
[2H]OS(=O)(=O)O[2H]
InChI
1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)/i/hD2
InChI key
QAOWNCQODCNURD-ZSJDYOACSA-N
Looking for similar products? Visit Product Comparison Guide
comparable product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?