Key Documents
520187
Sigma-Aldrich
Zirconium(IV) sulfate solution
35 wt. % in H2O, 99.9% trace metals basis (purity excludes Hf)
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
Zr(SO4)2
CAS Number:
Molecular Weight:
283.35
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
99.9% trace metals basis (purity excludes Hf)
form:
liquid
solubility:
H2O: slightly soluble(lit.)
ethanol: slightly soluble(lit.)
assay
99.9% trace metals basis (purity excludes Hf)
form
liquid
concentration
35 wt. % in H2O
solubility
H2O: slightly soluble(lit.)
ethanol: slightly soluble(lit.)
SMILES string
[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
InChI
1S/2H2O4S.Zr/c2*1-5(2,3)4;/h2*(H2,1,2,3,4);/q;;+4/p-4
InChI key
ZXAUZSQITFJWPS-UHFFFAOYSA-J
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
Reviews
No rating value
Active Filters