Key Documents
516252
Sigma-Aldrich
Rhodium(IV) oxide
99.9%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
RhO2
CAS Number:
Molecular Weight:
134.90
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay
99.9%
SMILES string
O=[Rh]=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)