Key Documents
498319
Sigma-Aldrich
4-(Phenylsulfonyl)-2-azetidinone
95%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C9H9NO3S
CAS Number:
Molecular Weight:
211.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
95%
mp
150-155 °C (lit.)
SMILES string
O=C1CC(N1)S(=O)(=O)c2ccccc2
pictograms
Corrosion
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)