Key Documents
497002
Sigma-Aldrich
4-O-Benzyl-D-glucal
97%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C13H16O4
CAS Number:
Molecular Weight:
236.26
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22
assay
97%
optical activity
[α]20/D +10°, c = 1 in chloroform
mp
99-103 °C (lit.)
SMILES string
C[C@H]1OC=C[C@@H](O)[C@@H]1OCc2ccccc2
InChI
1S/C13H16O4/c14-8-12-13(11(15)6-7-16-12)17-9-10-4-2-1-3-5-10/h1-7,11-15H,8-9H2/t11-,12-,13+/m1/s1
InChI key
OIWQZGBSQDJVJN-UPJWGTAASA-N
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
Reviews
No rating value
Active Filters