Key Documents
480703
Sigma-Aldrich
6-O-(tert-Butyldimethylsilyl)-D-galactal cyclic carbonate
97%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C13H22O5Si
CAS Number:
Molecular Weight:
286.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
assay
97%
optical activity
[α]20/D −66°, c = 2 in chloroform
mp
45-49 °C (lit.)
SMILES string
CC(C)(C)[Si](C)(C)OC[C@H]1OC=C[C@H]2OC(=O)O[C@@H]12
InChI
1S/C13H24O7Si/c1-13(2,3)21(4,5)18-7-8(15)11-10(16)9(6-14)19-12(17)20-11/h6,8-11,15-16H,7H2,1-5H3/t8-,9-,10-,11+/m1/s1
InChI key
AEXMDEVMJUZKHX-DBIOUOCHSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Important building block for both solution- and solid-phase synthesis of oligosaccharides.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)