Key Documents
478539
Sigma-Aldrich
2,4-Dimethylphenyl isocyanate
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(CH3)2C6H3NCO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
98%
refractive index
n20/D 1.534 (lit.)
bp
115 °C/2 mmHg (lit.)
density
1.041 g/mL at 25 °C (lit.)
SMILES string
Cc1ccc(N=C=O)c(C)c1
InChI
1S/C9H9NO/c1-7-3-4-9(10-6-11)8(2)5-7/h3-5H,1-2H3
InChI key
QUOBVYPFBJUOAJ-UHFFFAOYSA-N
pictograms
Health hazardExclamation mark
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
190.4 °F - closed cup
flash_point_c
88 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)