Key Documents
477621
Sigma-Aldrich
PSS-Butyronitrile-Heptacyclopentyl substituted
98%
Synonym(s):
3,5,7,9,11,13,15-Heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane-1-butyronitrile, 3-Cyanopropyl-POSS®
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C39H69NO12Si8
CAS Number:
Molecular Weight:
968.65
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
assay
98%
mp
300 °C (dec.) (lit.)
SMILES string
N#CCCC[Si]12O[Si]3(O[Si]4(O[Si](O1)(O[Si]5(O[Si](O2)(O[Si](O3)(O[Si](O4)(O5)C6CCCC6)C7CCCC7)C8CCCC8)C9CCCC9)C%10CCCC%10)C%11CCCC%11)C%12CCCC%12
Legal Information
POSS is a registered trademark of Hybrid Plastics, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)