Key Documents
477583
Sigma-Aldrich
2-Methoxybenzyl chloride
98%
Synonym(s):
2-(Chloromethyl)anisole
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3OC6H4CH2Cl
CAS Number:
Molecular Weight:
156.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
98%
contains
~0.3% propylene oxide as stabilizer
refractive index
n20/D 1.549 (lit.)
bp
122-124 °C/26 mmHg (lit.)
mp
28.3-29.8 °C (lit.)
density
1.125 g/mL at 25 °C (lit.)
SMILES string
COc1ccccc1CCl
pictograms
Health hazardCorrosion
signalword
Danger
Hazard Classifications
Carc. 1B - Eye Dam. 1 - Muta. 1B - Skin Corr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
210.2 °F - closed cup
flash_point_c
99 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)