Key Documents
477311
Sigma-Aldrich
(S)-(−)-1,2-Epithiooctane
90%
Synonym(s):
(S)-(−)-Hexylthiirane
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H16S
CAS Number:
Molecular Weight:
144.28
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
assay
90%
optical activity
[α]25/D −66°, neat
refractive index
n20/D 1.47 (lit.)
bp
83 °C/5 mmHg (lit.)
density
0.88 g/mL at 25 °C (lit.)
SMILES string
CCCCCC[C@H]1CS1
Choose from one of the most recent versions:
Certificates of Analysis (COA)