Key Documents
474444
Sigma-Aldrich
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-Eicosafluoroundecyl acrylate
97%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
H2C=CHCO2CH2(CF2)9CHF2
CAS Number:
Molecular Weight:
586.16
EC Number:
MDL number:
UNSPSC Code:
12162002
assay
97%
contains
0.01% 4-tert-butylcatechol as inhibitor
bp
98-101 °C/1.7 mmHg (lit.)
mp
45-53 °C (lit.)
SMILES string
FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)COC(=O)C=C
Choose from one of the most recent versions:
Certificates of Analysis (COA)