Key Documents
474061
Sigma-Aldrich
(2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoroheptyl)oxirane
97%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C9H5F13O
CAS Number:
Molecular Weight:
376.11
EC Number:
MDL number:
UNSPSC Code:
12162002
assay
97%
refractive index
n20/D 1.381 (lit.)
bp
80 °C/41 mmHg (lit.)
density
1.637 g/mL at 25 °C (lit.)
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC1CO1
Choose from one of the most recent versions:
Certificates of Analysis (COA)