Key Documents
474002
Sigma-Aldrich
(S)-(−)-4-(Methoxymethyl)-1,3-dioxolan-2-one
98%
Synonym(s):
(S)-3-Methoxy-1,2-propanediol cyclic carbonate
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C5H8O4
CAS Number:
Molecular Weight:
132.11
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
assay
98%
optical activity
[α]20/D −44°, neat
optical purity
ee: 97% (GLC)
refractive index
n20/D 1.437 (lit.)
bp
105-106 °C/0.9 mmHg (lit.)
density
1.24 g/mL at 25 °C (lit.)
SMILES string
COC[C@H]1COC(=O)O1
Choose from one of the most recent versions:
Certificates of Analysis (COA)