Key Documents
473650
Sigma-Aldrich
Glycidyl isobutyl ether
97%
Synonym(s):
(Isobutoxymethyl)oxirane, 2-(Isobutoxymethyl)oxirane, 2-[(2-Methylpropoxy)methyl]oxirane, Glycidyl isobutyl ether, Isobutyl glycidyl ether
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C7H14O2
CAS Number:
Molecular Weight:
130.18
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
assay
97%
refractive index
n20/D 1.416 (lit.)
bp
160 °C/750 mmHg (lit.)
density
0.909 g/mL at 25 °C (lit.)
SMILES string
CC(C)COCC1CO1
InChI
1S/C7H14O2/c1-6(2)3-8-4-7-5-9-7/h6-7H,3-5H2,1-2H3
InChI key
AQKDMKKMCVJJTC-UHFFFAOYSA-N
pictograms
FlameExclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
120.2 °F - closed cup
flash_point_c
49 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)