Key Documents
472913
Sigma-Aldrich
3,3′-Bithiophene
97%
Synonym(s):
3,3′-Dithienyl
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H6S2
CAS Number:
Molecular Weight:
166.26
MDL number:
UNSPSC Code:
12352100
assay
97%
mp
132-135 °C (lit.)
SMILES string
c1cc(cs1)-c2ccsc2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)