Key Documents
471879
Sigma-Aldrich
(1S)-(−)-(1-Amino-2-methylpropyl)phosphonic acid
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(CH3)2CHCH(NH2)P(O)(OH)2
CAS Number:
Molecular Weight:
153.12
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
assay
98%
optical activity
[α]20/D −1.0°, c = 1 in 1 M NaOH
mp
272-277 °C (lit.)
SMILES string
CC(C)[C@@H](N)P(O)(O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)