Key Documents
470996
Sigma-Aldrich
Lignin, alkali, carboxylated
average Mw ~175,000, average Mn ~16,000
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12162002
mol wt
average Mn ~16,000
average Mw ~175,000
extent of labeling
16 wt. % carboxylation
6 wt. % methoxy group
impurities
1 wt. % sulfur (nonsulfate)
pH
8 (3 wt. %)
solubility
H2O: soluble
Application
Complexing agent in water treatment and dispersing agent for dyestuffs.
Features and Benefits
Compatible with anionic and nonionic surfactants and wetting agents.
Choose from one of the most recent versions:
Certificates of Analysis (COA)