Key Documents
469149
Sigma-Aldrich
cis-Dichlorobis(dimethylphenylphosphine)palladium(II)
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
[C6H5P(CH3)2]2PdCl2
CAS Number:
Molecular Weight:
453.62
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
mp
185-187 °C (lit.)
SMILES string
Cl[Pd]Cl.CP(C)c1ccccc1.CP(C)c2ccccc2
Choose from one of the most recent versions:
Certificates of Analysis (COA)