Key Documents
462985
Sigma-Aldrich
2-Diazo-1-naphthol-5-sulfonic acid sodium salt monohydrate
technical grade
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H5N2NaO4S · H2O
CAS Number:
Molecular Weight:
290.23
EC Number:
UNSPSC Code:
12162002
grade
technical grade
mp
143 °C (dec.) (lit.)
SMILES string
O.[Na+].[O-]C1C(C=Cc2c1cccc2S([O-])(=O)=O)[N+]#N
pictograms
FlameExclamation mark
signalword
Danger
Hazard Classifications
Eye Irrit. 2 - Self-react. D - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
5.2 - Organic peroxides and self-reacting hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)