Key Documents
450707
Sigma-Aldrich
(R)-(−)-5,5-Dimethyl-4-phenyl-2-oxazolidinone
98%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C11H13NO2
CAS Number:
Molecular Weight:
191.23
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
assay
98%
optical activity
[α]25/D −71°, c = 2 in chloroform
optical purity
ee: 99% (HPLC)
mp
151-156 °C (lit.)
SMILES string
CC1(C)OC(=O)N[C@@H]1c2ccccc2
InChI
1S/C11H13NO2/c1-11(2)9(12-10(13)14-11)8-6-4-3-5-7-8/h3-7,9H,1-2H3,(H,12,13)/t9-/m1/s1
InChI key
HSQRCAULDOQKPF-SECBINFHSA-N
Application
Versatile chiral auxiliary for asymmetric synthesis used in diastereoselective Michael additions.[1]
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Davies, S.G. Sanganee, H.J.
Tetrahedron Asymmetry, 6, 671-671 (1995)