Key Documents
449369
Sigma-Aldrich
Ethyl 1-piperidinecarboxylate
97%
Synonym(s):
NSC 100911, NSC 236
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H15NO2
CAS Number:
Molecular Weight:
157.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
refractive index
n20/D 1.459 (lit.)
bp
183 °C (lit.)
density
1.018 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)N1CCCCC1
InChI
1S/C8H15NO2/c1-2-11-8(10)9-6-4-3-5-7-9/h2-7H2,1H3
InChI key
YSPVHAUJXLGZHP-UHFFFAOYSA-N
Application
Reactant for:
Electosynthesis of N-acyliminium ion equivalents using silica gel-supported piperidine
Application for anodic methoxylation of organic compounds
Identification of tetrahedral hemiacetal alkoxide intermediate
Carbonyl-alkene exchange reactions
Preparation of symmetrical ketones from organolithiums
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
186.8 °F - closed cup
flash_point_c
86 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Michiel J Thiecke et al.
Cell reports, 32(3), 107929-107929 (2020-07-23)
It is currently assumed that 3D chromosomal organization plays a central role in transcriptional control. However, depletion of cohesin and CTCF affects the steady-state levels of only a minority of transcripts. Here, we use high-resolution Capture Hi-C to interrogate the