Key Documents
448567
Sigma-Aldrich
(−)-α-Cubebene
97%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein/REAXYS Number:
6857364
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
assay
97%
optical activity
[α]20/D −23°, neat
contains
hydroquinone as stabilizer
refractive index
n20/D 1.481 (lit.)
bp
245-246 °C (lit.)
density
0.889 g/mL at 20 °C (lit.)
SMILES string
[H][C@]12[C@@H](CC[C@@H](C)[C@]13CC=C(C)[C@@H]23)C(C)C
Looking for similar products? Visit Product Comparison Guide
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Choose from one of the most recent versions:
Certificates of Analysis (COA)