Key Documents
447366
Sigma-Aldrich
(2-Furanylmethylene)malononitrile
98%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H4N2O
CAS Number:
Molecular Weight:
144.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
98%
mp
70-74 °C (lit.)
SMILES string
N#C\C(=C/c1ccco1)C#N
Gene Information
human ... EGFR(1956)
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)