Key Documents
446912
Sigma-Aldrich
1H-Benzotriazole-1-carboxaldehyde
90%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C7H5N3O
CAS Number:
Molecular Weight:
147.13
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
assay
90%
mp
90-95 °C (lit.)
SMILES string
O=Cn1nnc2ccccc12
Warning
Extremely hygroscopic
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Rémi Zallot et al.
The Biochemical journal, 463(1), 145-155 (2014-07-12)
The TenA protein family occurs in prokaryotes, plants and fungi; it has two subfamilies, one (TenA_C) having an active-site cysteine, the other (TenA_E) not. TenA_C proteins participate in thiamin salvage by hydrolysing the thiamin breakdown product amino-HMP (4-amino-5-aminomethyl-2-methylpyrimidine) to HMP