Key Documents
434329
Sigma-Aldrich
Silver(I) permanganate
Synonym(s):
Silver permanganate
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
AgMnO4
CAS Number:
Molecular Weight:
226.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
reaction suitability
reagent type: oxidant
SMILES string
[Ag].O[Mn](=O)(=O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)