Key Documents
430501
Sigma-Aldrich
Poly(1-vinylpyrrolidone)-graft-(1-hexadecene)
average Mn ~7,300
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12162002
mol wt
average Mn ~7,300
impurities
2-3% Nitrogen
refractive index
n20/D 1.479 (lit.)
relative viscosity
1.03-1.05, 1 wt. % in toluene(25 °C, Cannon-Fenske #100)(lit.)
density
0.9 g/mL at 25 °C (lit.)
General description
Film former with surface activity, water resistance and barrier properties.
Application
Pigment dispersant and wetting agent, substantive to skin.
pictograms
Exclamation mark
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)