Key Documents
429708
Sigma-Aldrich
o-Tolualdehyde 2,4-dinitrophenylhydrazone
environmental standard, 99%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3C6H4CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
300.27
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
grade
environmental standard
assay
99%
mp
195-198 °C (lit.)
SMILES string
Cc1ccccc1\C=N\Nc2ccc(cc2[N+]([O-])=O)[N+]([O-])=O
InChI
1S/C14H12N4O4/c1-10-4-2-3-5-11(10)9-15-16-13-7-6-12(17(19)20)8-14(13)18(21)22/h2-9,16H,1H3/b15-9+
InChI key
QUIFNJPMGQCNMH-OQLLNIDSSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)