Key Documents
427527
Sigma-Aldrich
Methyl 2-[3-(2-chloroethyl)ureido]benzoate
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
ClCH2CH2NHCONHC6H4CO2CH3
CAS Number:
Molecular Weight:
256.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
98%
mp
142 °C (lit.)
SMILES string
COC(=O)c1ccccc1NC(=O)NCCCl
InChI
1S/C11H13ClN2O3/c1-17-10(15)8-4-2-3-5-9(8)14-11(16)13-7-6-12/h2-5H,6-7H2,1H3,(H2,13,14,16)
InChI key
NUKAPQXOPXRWBX-UHFFFAOYSA-N
General description
Methyl 2-[3-(2-chloroethyl)ureido]benzoate is a compound that contains a ureido group (–NH–(C = O)–NH–).[1]
Application
Methyl 2-[3-(2-chloroethyl)ureido]benzoate was used as a model compound in assigning a possible structure to the nitrogen-rich segment in a fulvic acid (FA) from Pony Lake in Antarctica based on the 15N chemical-shift trends and 15N NMR.[1]
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
15N and 13C {14N} NMR investigation of the major nitrogen-containing segment in an aquatic fulvic acid: Evidence for a hydantoin derivative.
Fang X, et al.
Magnetic Resonance in Chemistry, 49(12), 775-780 (2011)