Key Documents
427225
Sigma-Aldrich
Poly(ethylene-alt-chlorotrifluoroethylene)
melt index 15-20 g/10 min (275 °C/2.16kg), pellets
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
[CH2CH2-CF(Cl)CF2]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
abrasion resistance
0.002 mL (Taber, 500 rev, ASTM D 1044)
form
pellets
melt index
15-20 g/10 min (275 °C/2.16kg)
dielectric constant
2.47, 1 kHz (ASTM D 150)
hardness
90 (Rockwell R, ASTM D 785)
transition temp
brittleness temperature -76 °C (ASTM D 746)
Tm 224 °C (DSC)
density
1.68 g/mL at 25 °C (lit.)
SMILES string
C=C.F\C(F)=C(\F)Cl
Looking for similar products? Visit Product Comparison Guide
Application
Wire and cable insulation, containers and seals for corrosive chemicals and liner for irradiation chambers and sterilizable tools.
Features and Benefits
No known solvent at temperatures < 120°C. Excellent chemical resistance, good electrical properties. Use range from cryogenic to 150°C. Excellent impact strength and abrasion resistance and resistant to radiation degradation. Crystallinity 50%.
Physical form
Crystalline, thermoplastic copolymer.
Choose from one of the most recent versions:
Certificates of Analysis (COA)