Key Documents
426334
Sigma-Aldrich
Methyl ethyl(tributylstannyl)carbamate
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
[CH3(CH2)3]3SnN(C2H5)CO2CH3
CAS Number:
Molecular Weight:
392.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
refractive index
n20/D 1.479 (lit.)
bp
90 °C/0.05 mmHg (lit.)
density
1.143 g/mL at 25 °C (lit.)
SMILES string
CCCC[Sn](CCCC)(CCCC)N(CC)C(=O)OC
Choose from one of the most recent versions:
Certificates of Analysis (COA)