Key Documents
422088
Sigma-Aldrich
CORFREE® M1
Synonym(s):
Dodecanedioic acid and sebacic acid, Mixture of undecanedioic acid
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
HO2C(CH2)nCO2H (n=8,9,10)
CAS Number:
Molecular Weight:
202.25
EC Number:
UNSPSC Code:
12352100
PubChem Substance ID:
mp
85-95 °C (lit.)
density
1.02 g/mL at 25 °C (lit.)
SMILES string
OC(=O)CCCCCCCCC(O)=O.OC(=O)CCCCCCCCCC(O)=O.OC(=O)CCCCCCCCCCC(O)=O
Legal Information
CORFREE is a registered trademark of Invista North America S.a.r.l.
Choose from one of the most recent versions:
Certificates of Analysis (COA)