Key Documents
416983
Sigma-Aldrich
Potassium thiosulfate hydrate
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
K2S2O3 · xH2O
Molecular Weight:
190.32 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form
solid
composition
Degree of hydration, ~0.5
SMILES string
[K+].[K+].[H]O[H].[O-]S([O-])(=O)=S
InChI
1S/2K.H2O3S2.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2
InChI key
RCEMYQVDTALKQN-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
pictograms
Exclamation mark
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)