Key Documents
414379
Sigma-Aldrich
Nickel(II) 2,9,16,23-tetraphenoxy-29H,31H-phthalocyanine
Dye content 97 %
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C56H32N8NiO4
CAS Number:
Molecular Weight:
939.60
MDL number:
UNSPSC Code:
12352103
composition
Dye content, 97%
λmax
678 nm
SMILES string
O(c1ccccc1)c2ccc3c4nc(nc5n6[Ni]n7c(nc8nc(nc6c9cc(Oc%10ccccc%10)ccc59)c%11ccc(Oc%12ccccc%12)cc8%11)c%13ccc(Oc%14ccccc%14)cc%13c7n4)c3c2
pictograms
Health hazard
signalword
Danger
Hazard Classifications
Carc. 1B - Resp. Sens. 1 - Skin Sens. 1
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)