Key Documents
412309
Sigma-Aldrich
Di(ethylene glycol) ethyl ether methacrylate
95%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
H2C=C(CH3)CO2(CH2CH2O)2C2H5
CAS Number:
Molecular Weight:
202.25
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
assay
95%
contains
100 ppm monomethyl ether hydroquinone as inhibitor
refractive index
n20/D 1.44 (lit.)
density
1 g/mL at 25 °C (lit.)
SMILES string
CCOCCOCCOC(=O)C(C)=C
Choose from one of the most recent versions:
Certificates of Analysis (COA)