Key Documents
409111
Sigma-Aldrich
Poly(dimethylamine-co-epichlorohydrin) solution
50 wt. % in H2O
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
[N(CH3)2(Cl)CH2CH(OH)CH2]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
concentration
50 wt. % in H2O
refractive index
n20/D 1.427
density
1.1 g/mL at 25 °C
SMILES string
CNC.ClCC1CO1
Choose from one of the most recent versions:
Certificates of Analysis (COA)