Key Documents
408034
Sigma-Aldrich
Poly[(o-cresyl glycidyl ether)-co-formaldehyde]
average Mn ~1,270
Synonym(s):
Formaldehyde, polymer with (chloromethyl)oxirane and 2-methylphenol
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12162002
mol wt
average Mn ~1,270
extent of labeling
5.4 epoxide functionality
0.425 eq epoxy per 100 g
viscosity
60 cP(lit.)
mp
70-75 °C (lit.)
85-95 °C
transition temp
Tg 37-39 °C (DSC)
softening point 85-100 °C (ring and ball)
density
1.12 g/mL at 25 °C (lit.)
InChI
1S/C7H8O.C3H5ClO.CH2O/c1-6-4-2-3-5-7(6)8;4-1-3-2-5-3;1-2/h2-5,8H,1H3;3H,1-2H2;1H2
InChI key
XRYDVSRMOIMZGM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
For high temperature adhesives, coatings, electrical and laminating products.
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)