Key Documents
401250
Sigma-Aldrich
Lithium chromate hydrate
technical grade
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
Li2CrO4
CAS Number:
Molecular Weight:
129.88 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade
technical grade
SMILES string
[Li+].[Li+].O.[O-][Cr]([O-])(=O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)