Key Documents
401021
Sigma-Aldrich
Chromium(III) telluride
−325 mesh
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
Cr2Te3
CAS Number:
Molecular Weight:
486.79
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
form
powder
particle size
−325 mesh
SMILES string
[Te]=[Cr][Te][Cr]=[Te]
InChI
1S/2Cr.3Te
InChI key
PDJHBRMODJKXLQ-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)