Key Documents
400491
Sigma-Aldrich
Tellurium tetraiodide
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
TeI4
CAS Number:
Molecular Weight:
635.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
density
5.05 g/mL at 25 °C (lit.)
SMILES string
I[Te](I)(I)I
Choose from one of the most recent versions:
Certificates of Analysis (COA)