Key Documents
399183
Sigma-Aldrich
4′-Bromobenzo-18-crown-6
98%
Synonym(s):
(Benzo-18-crown-6)-4′-yl bromide
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C16H23BrO6
CAS Number:
Molecular Weight:
391.25
Beilstein/REAXYS Number:
4818529
MDL number:
UNSPSC Code:
12352103
assay
98%
mp
78-80 °C (lit.)
SMILES string
Brc1ccc2OCCOCCOCCOCCOCCOc2c1
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)