Key Documents
393010
Sigma-Aldrich
Coumarin 337
Dye content 99 %
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C16H14N2O2
CAS Number:
Molecular Weight:
266.29
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
composition
Dye content, 99%
mp
250 °C (dec.) (lit.)
λmax
443 nm
SMILES string
O=C1Oc2c3CCCN4CCCc(cc2C=C1C#N)c34
Application
Suitable as a laser dye
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)