Key Documents
389013
Sigma-Aldrich
Stearyl citrate
technical grade
Sign Into View Organizational & Contract Pricing
About This Item
CAS Number:
EC Number:
UNSPSC Code:
12352103
grade
technical grade
Choose from one of the most recent versions:
Certificates of Analysis (COA)