Key Documents
387835
Sigma-Aldrich
1-Deoxy-1-nitro-D-iditol hemihydrate
99%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C6H13NO7 · .5H2O
Molecular Weight:
220.18
UNSPSC Code:
12352200
PubChem Substance ID:
assay
99%
optical activity
[α]20/D +4.0°, c = 2 in H2O
mp
88 °C (lit.)
SMILES string
O.OC[C@@H](O)[C@H](O)[C@@H](O)[C@H](O)C[N+]([O-])=O.OC[C@@H](O)[C@H](O)[C@@H](O)[C@H](O)C[N+]([O-])=O
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)