Key Documents
383430
Sigma-Aldrich
6-Chlorochromone
97%
Synonym(s):
6-Chloro-1-benzopyran-4(4H)-one
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C9H5ClO2
CAS Number:
Molecular Weight:
180.59
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
assay
97%
mp
136-138 °C (lit.)
SMILES string
Clc1ccc2OC=CC(=O)c2c1
Choose from one of the most recent versions:
Certificates of Analysis (COA)