Key Documents
378747
Sigma-Aldrich
Dimethyl-1,1,1-d3 sulfide
99.5 atom % D
Synonym(s):
(Methyl sulfide)-1,1,1-d3
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
CH3SCD3
CAS Number:
Molecular Weight:
65.15
MDL number:
UNSPSC Code:
12142201
vapor pressure
8.02 psi ( 20 °C)
isotopic purity
99.5 atom % D
mol wt
65.16
refractive index
n20/D 1.433 (lit.)
bp
38 °C (lit.)
mp
-98 °C (lit.)
density
0.887 g/mL at 25 °C (lit.)
mass shift
M+3
SMILES string
[2H]C([2H])([2H])SC
InChI
1S/C2H6S/c1-3-2/h1-2H3/i1D3
InChI key
QMMFVYPAHWMCMS-FIBGUPNXSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)