Key Documents
369624
Sigma-Aldrich
Ethyl (S)-cis-3-(2,2-dimethyl-1,3-dioxolan-4-yl)propenoate
99%
Synonym(s):
Ethyl (S)-cis-4,5-O-isopropylidene-4,5-dihydroxy-2-pentenoate
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H16O4
CAS Number:
Molecular Weight:
200.23
Beilstein/REAXYS Number:
4231889
MDL number:
UNSPSC Code:
12352103
assay
99%
optical activity
[α]20/D +123°, c = 1 in chloroform
refractive index
n20/D 1.443 (lit.)
density
1.038 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)\C=C/[C@H]1COC(C)(C)O1
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)