Key Documents
363707
Sigma-Aldrich
Tetraethylammonium bis(acetonitrile)tetrachlororuthenate(III)
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
(C2H5)4N[(CH3CN)2RuCl4]
CAS Number:
Molecular Weight:
455.24
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
mp
220 °C (dec.) (lit.)
SMILES string
CC#N.CC#N.Cl[Ru-](Cl)(Cl)Cl.CC[N+](CC)(CC)CC
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.